9,9'-Spirobi[9H-fluorene] 98%

Supplier: Thermo Scientific Chemicals

H64133-03 H64133-06 H64133-MD
AAH64133-03EA 53.6 USD
AAH64133-03 AAH64133-06 AAH64133-MD
9,9'-Spirobi[9H-fluorene] 98%
9,9'-Spirobi[9H-fluorene]

Purity: 98% CAS No.: 159-66-0 Molecular Formula: C25H16 Formula Weight: 316.40 Physical Form: Solid Appearance: White to pale yellow Melting Point: 195-200° Beilstein Reference: 1914463 MDL No.: MFCD08704216

Formula: C₂₅H₁₆
Melting Pt: 195…200 °C
Storage Temperature: Ambient
MDL Number: MFCD08704216
CAS Number: 159-66-0

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR