6-Chloro-2-pyridinecarboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloropyridine-2-carboxaldehyde, 6-Chloro-2-formylpyridine, 6-Chloropicolinaldehyde

H64129-03 H64129-06 H64129-MD
AAH64129-03EA 113 USD
AAH64129-03 AAH64129-06 AAH64129-MD
6-Chloro-2-pyridinecarboxaldehyde 97%
6-Chloro-2-pyridinecarboxaldehyde

Purity: 97% CAS No.: 54087-03-5 Molecular Formula: C6H4ClNO Formula Weight: 141.55 Physical Form: Powder Appearance: White Melting Point: 69-70° Beilstein Reference: 1446583 MDL No.: MFCD09832941

Formula: C₆H₄ClNO
Melting Pt: 69…70 °C
Storage Temperature: Ambient
MDL Number: MFCD09832941
CAS Number: 54087-03-5

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR