5-Bromo-2-chloro-3-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64109-06
H64109-14
AAH64109-06EA
46.09
USD
AAH64109-06
AAH64109-14
5-Bromo-2-chloro-3-methylpyridine 98%
5-Bromo-2-chloro-3-methylpyridine
Purity: 98%
CAS No.: 29241-60-9
Molecular Formula: C6H5BrClN
Formula Weight: 206.47
Physical Form: Solid
Melting Point: 40-44°
Beilstein Reference: 1562796
MDL No.: MFCD03095093
Formula:
C₆H₅BrClN Melting Pt: 40…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095093 CAS Number: 29241-60-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...