Imidazole-4-carboxylic acid ethyl ester 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl imidazole-4-carboxylate
H64095-06
H64095-14
AAH64095-03EA
23.76
USD
AAH64095-03
AAH64095-06
AAH64095-14
Imidazole-4-carboxylic acid ethyl ester 98%
Imidazole-4-carboxylic acid ethyl ester
Purity: 98%
CAS No.: 23785-21-9
Molecular Formula: C6H8N2O2
Formula Weight: 140.14
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 156-160°
Beilstein Reference: 606937
MDL No.: MFCD02646639
Formula:
C₆H₈N₂O₂ MW: 140.14 g/mol Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD02646639 CAS Number: 23785-21-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...