2-Hydrazinopyrazine 98%

Supplier: Thermo Scientific Chemicals
Warning

H64086-03 H64086-06
AAH64086-03EA 57.5 USD
AAH64086-03 AAH64086-06 AAH64086-MD
2-Hydrazinopyrazine 98%
2-Hydrazinopyrazine

Purity: 98% CAS No.: 54608-52-5 Molecular Formula: C4H6N4 Formula Weight: 110.12 Physical Form: Solid Appearance: Yellow Melting Point: 109-113° Beilstein Reference: 606630 MDL No.: MFCD04114555

Formula: C4H6N4
MW: 110.12 g/mol
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD04114555
CAS Number: 54608-52-5

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR