Methyl-4,6-dichloronicotinate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 4,6-dichloronicotinate
H64077-06
H64077-14
AAH64077-03EA
25.1
USD
AAH64077-03
AAH64077-06
AAH64077-14
Methyl-4,6-dichloronicotinate 95%
Methyl-4,6-dichloronicotinate
Purity: 95%
CAS No.: 65973-52-6
Molecular Formula: C7H5Cl2NO2
Formula Weight: 206.02
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 13279782
MDL No.: MFCD04125732
Formula:
C₇H₅Cl₂NO₂ MW: 206.03 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04125732 CAS Number: 65973-52-6 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...