3-cyclopropyl-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning

H64069-03 H64069-06
AAH64069-03EA 93.2 USD
AAH64069-03 AAH64069-06 AAH64069-MD
3-cyclopropyl-1H-pyrazole 98%
3-cyclopropyl-1H-pyrazole

Purity: 98% CAS No.: 100114-57-6 Molecular Formula: C6H8N2 Formula Weight: 108.14 Physical Form: Liquid Appearance: Colorless Beilstein Reference: 1561659 MDL No.: MFCD04970938

Formula: C₆H₈N₂
Storage Temperature: Ambient
MDL Number: MFCD04970938
CAS Number: 100114-57-6

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR