9-Phenylcarbazole-3-boronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64059-03
H64059-06
AAH64059-03EA
62.2
USD
AAH64059-03
AAH64059-06
AAH64059-MD
9-Phenylcarbazole-3-boronic acid 98%
Purity: 98%
CAS No.: 854952-58-2
Molecular Formula: C18H14BNO2
Formula Weight: 287.13
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 10566519
MDL No.: MFCD12196936
Formula:
C₁₈H₁₄BNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD12196936 CAS Number: 854952-58-2 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...