trans-3-Chloro-β-nitrostyrene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64053-03
H64053-06
AAH64053-03EA
32.8
USD
AAH64053-03
AAH64053-06
trans-3-Chloro-β-nitrostyrene 97%
trans-3-Chloro-β-nitrostyrene
Purity: 97%
CAS No.: 37888-03-2
Molecular Formula: C8H6ClNO2
Formula Weight: 183.59
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 48-52°
Beilstein Reference: 2087804
MDL No.: MFCD00175518
Formula:
C₈H₆ClNO₂ Melting Pt: 48…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00175518 CAS Number: 37888-03-2 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...