4-Bromo-2,3-difluorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

H64044-03 H64044-06
AAH64044-03EA 69.7 USD
AAH64044-03 AAH64044-06 AAH64044-MD
4-Bromo-2,3-difluorobenzaldehyde 98%
4-Bromo-2,3-difluorobenzaldehyde

Purity: 98% CAS No.: 644985-24-0 Molecular Formula: C7H3BrF2O Formula Weight: 221.00 Physical Form: Powder Appearance: Off-white Melting Point: 66-67° Beilstein Reference: 9551272 MDL No.: MFCD09258953

Formula: C₇H₃BrF₂O
MW: 221.00 g/mol
Melting Pt: 66…67 °C
Storage Temperature: Ambient
MDL Number: MFCD09258953
CAS Number: 644985-24-0

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR