2-Bromo-m-toluidine 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64014-03
H64014-MD
AAH64014-03EA
89.4
USD
AAH64014-03
AAH64014-06
AAH64014-MD
2-Bromo-m-toluidine 97+%
2-Bromo-m-toluidine
Purity: 97+%
CAS No.: 54879-20-8
Molecular Formula: C7H8BrN
Formula Weight: 186.05
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₇H₈BrN MW: 186.05 g/mol Boiling Pt: 48 °C/0.03 mm Density: 1.489 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD06411367 CAS Number: 54879-20-8 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...