4-(Trifluoromethoxy)phenyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-(Trifluoromethoxy)phenylisothiocyanate
H64013-03
H64013-06
AAH64013-03EA
41.8
USD
AAH64013-03
AAH64013-06
4-(Trifluoromethoxy)phenyl isothiocyanate 97%
4-(Trifluoromethoxy)phenyl isothiocyanate
Purity: 97%
CAS No.: 64285-95-6
Molecular Formula: C8H4F3NOS
Formula Weight: 219.18
Storage Temperature: 2°C to 8°C
Flash Point: 107°(225°F)
Physical Form: Liquid
Density: 1.35
Refractive Index: 1.5410
Sensitivity: Moisture Sensitive
Appearance: Colorless
Boiling Point: 215-218° lit.
Beilstein Reference: 2723939
UN No.: 1760
MDL No.: MFCD00042412
Formula:
C₈H₄F₃NOS MW: 219.19 g/mol Boiling Pt: 215…218° (lit.) Density: 1.35 Flash Pt: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042412 CAS Number: 64285-95-6 UN: 1760 ADR: 8,III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...