3-Bromo-5-iodopyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64007-03
H64007-06
AAH64007-03EA
133
USD
AAH64007-03
AAH64007-06
AAH64007-MD
3-Bromo-5-iodopyridine 95%
3-Bromo-5-iodopyridine
Purity: 95%
CAS No.: 233770-01-9
Molecular Formula: C5H3BrIN
Formula Weight: 283.89
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
Melting Point: 127-131°
Beilstein Reference: 108478
MDL No.: MFCD03086019
Formula:
C₅H₃BrIN MW: 283.89 g/mol Melting Pt: 127…131 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03086019 CAS Number: 233770-01-9 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...