(1-Methyl-1H-imidazol-2-yl)methanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-2-imidazolemethanol

H64006-06 H64006-14
AAH64006-06EA 58.6 USD
AAH64006-06 AAH64006-14
(1-Methyl-1H-imidazol-2-yl)methanol 98%
(1-Methyl-1H-imidazol-2-yl)methanol

Purity: 98% CAS No.: 17334-08-6 Molecular Formula: C5H8N2O Formula Weight: 112.13 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 107-111° Beilstein Reference: 113492 MDL No.: MFCD00964673

Formula: C₅H₈N₂O
MW: 112.13 g/mol
Melting Pt: 107…111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00964673
CAS Number: 17334-08-6

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR