1H-Indazole-3-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Indazole-3-carboxylic acid
H64001-06
H64001-14
H64001-22
AAH64001-06EA
36.6
USD
AAH64001-06
AAH64001-14
AAH64001-22
1H-Indazole-3-carboxylic acid 98%
1H-Indazole-3-carboxylic acid
Purity: 98%
CAS No.: 4498-67-3
Molecular Formula: C8H6N2O2
Formula Weight: 162.15
Physical Form: Powder
Appearance: Pale brown
Melting Point: 266-270° lit.
Beilstein Reference: 6354
MDL No.: MFCD00211066
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Melting Pt: 266…270 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00211066 CAS Number: 4498-67-3 EINECS: 224-794-5 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...