7-Bromoquinoline 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63987-03
H63987-06
AAH63987-03EA
110
USD
AAH63987-03
AAH63987-06
AAH63987-MD
7-Bromoquinoline 95%
7-Bromoquinoline
Purity: 95%
CAS No.: 4965-36-0
Molecular Formula: C9H6BrN
Formula Weight: 208.06
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD03695823
Formula:
C9H6BrN Storage Temperature: Refrigerator |
MDL Number:
MFCD03695823 CAS Number: 4965-36-0 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...