N,N'-Dicarbobenzoxy-L-cystine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(Z-Cystine-OH)₂, (Z-Cys-OH)2, N,N'-Di-Cbz-L-cystine, N,N'-Dibenzyloxycarbonyl-L-cystine
H63917-06
H63917-14
AAH63917-03EA
15.37
USD
AAH63917-03
AAH63917-06
AAH63917-14
N,N'-Dicarbobenzoxy-L-cystine 98%
N,N'-Dicarbobenzoxy-L-cystine
Purity: 98%
CAS No.: 6968-11-2
Molecular Formula: C22H24N2O8S2
Formula Weight: 508.56
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 119°
Beilstein Reference: 2550640
MDL No.: MFCD00022030
Formula:
C₂₂H₂₄N₂O₈S₂ MW: 508.56 g/mol Melting Pt: 119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022030 CAS Number: 6968-11-2 EINECS: 230-181-3 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...