3-Methyl-5-isoxazolecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methylisoxazole-5-carboxylic acid

H63740-03 H63740-MD
AAH63740-03EA 249 USD
AAH63740-03 AAH63740-MD
3-Methyl-5-isoxazolecarboxylic acid 97%
3-Methyl-5-isoxazolecarboxylic acid

Purity: 97% CAS No.: 4857-42-5 Molecular Formula: C5H5NO3 Formula Weight: 127.10 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD00464222

Formula: C₅H₅NO₃
MW: 127.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00464222
CAS Number: 4857-42-5
EINECS: 225-454-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR