(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-Cys(Bzl)-OH, N-Boc-S-benzyl-L-cysteine
H63736-06
H63736-14
AAH63736-06EA
32.3
USD
AAH63736-06
AAH63736-14
AAH63736-22
(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%
(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid
Purity: 98%
CAS No.: 5068-28-0
Molecular Formula: C15H21NO4S
Formula Weight: 311.4
Physical Form: Solid
Melting Point: 86-88°
Beilstein Reference: 2000417
Optical Rotation: -44°(c=1 in acetic acid)
MDL No.: MFCD00065567
Formula:
C15H21NO4S MW: 311.4 g/mol Melting Pt: 86…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065567 CAS Number: 5068-28-0 EINECS: 225-772-8 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...