4-Bromophenethylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(4-Bromophenyl)ethylamine

H63706-03 H63706-06 H63706-14
AAH63706-03EA 47.5 USD
AAH63706-03 AAH63706-06 AAH63706-14
4-Bromophenethylamine 98%
4-Bromophenethylamine

Purity: 98% CAS No.: 73918-56-6 Molecular Formula: C8H10BrN Formula Weight: 200.08 Storage Temperature: 2°C to 8°C Flash Point: 113°(235°F) Physical Form: Liquid Density: 1.290 Refractive Index: 1.5740 Appearance: Colorless Boiling Point: 63-72°/0.2mm UN No.: 2735 MDL No.: MFCD00008189

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 63…72 °C/0.2 mm
Density: 1.290
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008189
CAS Number: 73918-56-6
EINECS: 277-636-2
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR