2-Boc-2-azabicyclo[2.2.1]hept-5-ene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
tert-Butyl 2-Azabicyclo[2.2.1]hept-5-ene-2-carboxylate, 2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]hept-5-ene, 2-Azabicyclo[2.2.1]hept-5-ene-2-carboxylic acid tert-Butyl ester, 2-Boc-2-azabicyclo[2.2.1]hept-5-ene
H63698-03
H63698-06
AAH63698-03EA
130
USD
AAH63698-03
AAH63698-06
AAH63698-MD
2-Boc-2-azabicyclo[2.2.1]hept-5-ene 98%
2-Boc-2-azabicyclo[2.2.1]hept-5-ene
Purity: 98%
CAS No.: 188345-71-3
Molecular Formula: C11H17NO2
Formula Weight: 195.26
Physical Form: Liquid
Appearance: Yellow
MDL No.: MFCD22416354
Formula:
C₁₁H₁₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD22416354 CAS Number: 188345-71-3 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...