7-Chloro-1,2,3,4-tetrahydroisoquinoline 95%

Supplier: Thermo Scientific Chemicals
Warning

H63693-03 H63693-MD
AAH63693-03EA 626 USD
AAH63693-03 AAH63693-MD
7-Chloro-1,2,3,4-tetrahydroisoquinoline 95%
7-Chloro-1,2,3,4-tetrahydroisoquinoline

Purity: 95% CAS No.: 82771-60-6 Molecular Formula: C9H10ClN Formula Weight: 167.64 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD05861543

Formula: C₉H₁₀ClN
MW: 167.636 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05861543
CAS Number: 82771-60-6

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR