8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-NH-PEG2-CH2COOH, 8-(Fmoc-amino)-3,6-dioxa-n-octanoic acid, Fmoc-8-amino-3,6-dioxaoctanoic acid, [2-[2-[(Fmoc-amino)ethoxy]ethoxy]acetic acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, [2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]acetic acid
H63641-03
H63641-MD
AAH63641-03EA
329
USD
AAH63641-03
AAH63641-MD
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid 95%
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid
Purity: 95%
CAS No.: 166108-71-0
Molecular Formula: C21H23NO6
Formula Weight: 385.42
Physical Form: Crystalline powder
Appearance: White
Beilstein Reference: 9658732
MDL No.: MFCD01321015
Formula:
C₂₁H₂₃NO₆ MW: 385.42 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01321015 CAS Number: 166108-71-0 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...