2-Bromo-3-fluorobenzyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromo-1-(bromomethyl)-3-fluorobenzene

H63632-03 H63632-06
AAH63632-03EA 47.7 USD
AAH63632-03 AAH63632-06
2-Bromo-3-fluorobenzyl bromide 98%
2-Bromo-3-fluorobenzyl bromide

Purity: 98% CAS No.: 1184918-22-6 Molecular Formula: C7H5Br2F Formula Weight: 267.92 UN No.: 3261 MDL No.: MFCD12911576

Formula: C₇H₅Br₂F
Storage Temperature: Ambient
MDL Number: MFCD12911576
CAS Number: 1184918-22-6
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR