Boc-Lys(Z)-OSu 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Nepsilon-Benzyloxycarbonyl-Nα-Boc-L-lysine N-succinimidyl ester

H63470-06 H63470-14
AAH63470-03EA 95.84 USD
AAH63470-03 AAH63470-06 AAH63470-14
Boc-Lys(Z)-OSu 95%
Boc-Lys(Z)-OSu

Purity: 95% CAS No.: 34404-36-9 Molecular Formula: C23H31N3O8 Formula Weight: 477.51 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Melting Point: 108-117° Beilstein Reference: 1559393 Optical Rotation: -21° (c=2 in dioxane) MDL No.: MFCD00037915

Formula: C₂₃H₃₁N₃O₈
Melting Pt: 108…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037915
CAS Number: 34404-36-9
EINECS: 251-999-7

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR