2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63447-03
H63447-MD
AAH63447-03EA
200
USD
AAH63447-03
AAH63447-06
AAH63447-MD
2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone 95%
2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinone
Purity: 95%
CAS No.: 315-08-2
Molecular Formula: C10H6F3NOS2
Formula Weight: 277.28
Physical Form: Solid
Appearance: Off-white
MDL No.: MFCD00126176
Formula:
C₁₀H₆F₃NOS₂ Storage Temperature: Ambient |
MDL Number:
MFCD00126176 CAS Number: 315-08-2 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...