2,4-Dichloro-3-cyanopyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Dichloropyridine-3-carbonitrile, 2,4-Dichloronicotinonitrile

H63347-03 H63347-06
AAH63347-03EA 84.9 USD
AAH63347-03 AAH63347-06 AAH63347-MD
2,4-Dichloro-3-cyanopyridine 95%
2,4-Dichloro-3-cyanopyridine

Purity: 95% CAS No.: 180995-12-4 Molecular Formula: C6H2Cl2N2 Formula Weight: 173.00 Physical Form: Solid UN No.: 3439 MDL No.: MFCD09750302

Formula: C₆H₂Cl₂N₂
MW: 172.996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750302
CAS Number: 180995-12-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR