(2-Chloro-3-pyridinyl)methanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-3-pyridinemethanol, (2-chloropyridin-3-yl)methanol, 2-Chloro-3-pyridinylmethanol
H63341-03
H63341-06
AAH63341-03EA
45.6
USD
AAH63341-03
AAH63341-06
(2-Chloro-3-pyridinyl)methanol 97%
(2-Chloro-3-pyridinyl)methanol
Purity: 97%
CAS No.: 42330-59-6
Molecular Formula: C6H6ClNO
Formula Weight: 143.57
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 61-65° dec
Boiling Point: 112°/0.2mm
Beilstein Reference: 1525378
MDL No.: MFCD01935961
Formula:
C₆H₆ClNO MW: 143.57 g/mol Boiling Pt: 112 °C/0.2 mm Melting Pt: 61…65 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01935961 CAS Number: 42330-59-6 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...