Thiazole-2-carbaldehyde 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Thiazolecarboxaldehyde, Thiazole-2-carboxaldehyde

H63329-03 H63329-06
AAH63329-03EA 47.3 USD
AAH63329-03 AAH63329-06
Thiazole-2-carbaldehyde 95%
Thiazole-2-carbaldehyde

Purity: 95% CAS No.: 10200-59-6 Molecular Formula: C4H3NOS Formula Weight: 113.13 Storage Temperature: 2°C to 8°C Flash Point: 68°(154°F) Physical Form: Liquid Density: 1.288 Refractive Index: 1.574 Appearance: Colorless to pale yellow Boiling Point: 61-63°/15mm MDL No.: MFCD00142924

Formula: C₄H₃NOS
Boiling Pt: 61…63 °C/15 mm
Density: 1.288
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00142924
CAS Number: 10200-59-6

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR