Z-Dab(Boc)-OH·DCHA 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt

H62772-03 H62772-06
AAH62772-03EA 77.6 USD
AAH62772-03 AAH62772-06 AAH62772-MD
Z-Dab(Boc)-OH·DCHA 98%
Z-Dab(Boc)-OH·DCHA

Purity: 98% CAS No.: 3350-13-8 Molecular Formula: C17H24N2O6.C12H23N Formula Weight: 553.70 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white MDL No.: MFCD00237341

Formula: C₂₉H₄₇N₃O₆
MW: 533.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237341
CAS Number: 3350-13-8

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR