2-Amino-5-fluorobenzeneboronic acidpinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5-fluorophenyl boronic acid pinacol ester, 4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

H62575-03 H62575-MD
AAH62575-03EA 333 USD
AAH62575-03 AAH62575-MD
2-Amino-5-fluorobenzeneboronic acidpinacol ester 96%
2-Amino-5-fluorobenzeneboronic acidpinacol ester

Purity: 96% CAS No.: 863578-24-9 Molecular Formula: C12H17BFNO2 Formula Weight: 237.08 Physical Form: Solid Appearance: Off-white Melting Point: 49-50° MDL No.: MFCD11617916

Formula: C₁₂H₁₇BFNO₂
MW: 237.081 g/mol
Melting Pt: 49…50 °C
Storage Temperature: Ambient
MDL Number: MFCD11617916
CAS Number: 863578-24-9

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR