Boc-Hyp-OBZL 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-trans-4-hydroxy-L-proline benzyl ester, (2S,4R)-2-Benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate
H62495-03
H62495-06
AAH62495-03EA
95.1
USD
AAH62495-03
AAH62495-06
AAH62495-MD
Boc-Hyp-OBZL 95%
Boc-Hyp-OBZL
Purity: 95%
CAS No.: 89813-47-8
Molecular Formula: C17H23NO5
Formula Weight: 321.37
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Pale brown
MDL No.: MFCD00076979
Formula:
C₁₇H₂₃NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00076979 CAS Number: 89813-47-8 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...