Thiophene-3-methylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Thienylmethylamine
H61487-03
H61487-06
AAH61487-03EA
156
USD
AAH61487-03
AAH61487-06
Thiophene-3-methylamine 97%
Thiophene-3-methylamine
Purity: 97%
CAS No.: 27757-86-4
Molecular Formula: C5H7NS
Formula Weight: 113.18
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5660
Boiling Point: 102°/15mm
UN No.: 2735
MDL No.: MFCD01529872
Formula:
C₅H₇NS Boiling Pt: 102 °C/15 mm Density: 1.130 Storage Temperature: Ambient |
MDL Number:
MFCD01529872 CAS Number: 27757-86-4 UN: 2735 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...