4,6-Dichloronicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H61381-06 H61381-14
AAH61381-06EA 129 USD
AAH61381-06 AAH61381-14
4,6-Dichloronicotinic acid 97%
4,6-Dichloronicotinic acid

Purity: 97% CAS No.: 73027-79-9 Molecular Formula: C6H3Cl2NO2 Formula Weight: 192.00 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: White MDL No.: MFCD00234146

Formula: C₆H₃Cl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00234146
CAS Number: 73027-79-9

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR