tert-Butyl-3-bromopyrrolidine-1-carboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

H57071-06 H57071-03
AAH57071-06EA 509 USD
AAH57071-06 AAH57071-03 AAH57071-MD
tert-Butyl-3-bromopyrrolidine-1-carboxylate 95%
tert-Butyl-3-bromopyrrolidine-1-carboxylate

Purity: 95% CAS No.: 939793-16-5 Molecular Formula: C9H16BrNO2 Formula Weight: 250.14 Storage Temperature: 2°C to 8°C Melting Point: 45-53° UN No.: 3077 MDL No.: MFCD08752478

Formula: C9H16BrNO2
Melting Pt: 45…53 °C
Storage Temperature: Ambient
MDL Number: MFCD08752478
CAS Number: 939793-16-5
UN: 3077
ADR: "9",III

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR