1-Phenyl-2-(trimethylsilyl)acetylene 99%

Supplier: Thermo Scientific Chemicals
Warning

H55866-06 H55866-14 H55866-22
AAH55866-06EA 39.6 USD
AAH55866-06 AAH55866-14 AAH55866-22
1-Phenyl-2-(trimethylsilyl)acetylene 99%
1-Phenyl-2-(trimethylsilyl)acetylene

Purity: 99% CAS No.: 2170-06-1 Molecular Formula: C11H14Si Formula Weight: 174.32 Flash Point: 74°(165°F) Physical Form: Liquid Density: 0.886 Refractive Index: 1.5280 Appearance: Colorless Boiling Point: 87-88°/9mm Beilstein Reference: 3088678 MDL No.: MFCD00054860

Formula: C₁₁H₁₄Si
Boiling Pt: 87…88 °C/9 mm
Density: 0.886
Flash Pt: 74 °C(165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00054860
CAS Number: 2170-06-1

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR