(4-Bromophenylethynyl)trimethylsilane 98%

Supplier: Thermo Scientific Chemicals

Synonyms: ((4-Bromophenyl)ethynyl)trimethylsilane

H55534-03 H55534-06 H55534-14
AAH55534-03EA 33.6 USD
AAH55534-03 AAH55534-06 AAH55534-14
(4-Bromophenylethynyl)trimethylsilane 98%
(4-Bromophenylethynyl)trimethylsilane

Purity: 98% CAS No.: 16116-78-2 Molecular Formula: C11H13BrSi Formula Weight: 253.21 Physical Form: Powder Appearance: White to yellow Melting Point: 61-63° MDL No.: MFCD01863556

Formula: C₁₁H₁₃BrSi
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD01863556
CAS Number: 16116-78-2

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR