Pin Spanner Wrench for 58 - 62 mm Nuts, Cytiva
Supplier: Cytiva
28964776 28964777 28964778
101725-276EA 160.31 USD
101725-276 104013-122 104013-124
Pin Spanner Wrench for 58 - 62 mm Nuts, Cytiva
Wrenches
These spanner wrenches are used with HiScale™ columns. To readjust the position of the adapter in the column a spanner can be used to turn the end cap. To prevent the column from rotating, a second spanner can be used to hold the end housing.