Packing Connectors, Cytiva
Supplier: Cytiva
18115321 18115323 18115322
101724-864EA 134.46 USD
101724-864 101724-868 101724-866
Packing Connectors, Cytiva
Fittings and Connectors
Packing Tricorn columns is simplified using Tricorn packing connector and the appropriate glass tube.
Tricorn packing connector includes one packing connector and two EPDM O-rings. Packing connectors are available in three sizes for using different i.d. tubes: 5-5 is for 5 mm tubes on both sides; 5-10 is for a 5 mm tube on one side and a 10 mm tube on the other side; and 10-10 is for 10 mm tubes on both sides. Ethylene propylene diene change characteristics when exposed to some organic chemicals. For repetitive long term use, use per fluor rubber (PFR).