1,1,1-Trifluoroacetylacetone ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,1,1-Trifluoropentane-2,4-dione, 1,1,1-Trifluoro-2,4-pentanedione
225875-100G
225875-25G
225875-5G
BT225875-100GEA
298.46
USD
BT225875-100G
BT225875-25G
BT225875-5G
1,1,1-Trifluoroacetylacetone ≥98%
1,1,1-Trifluoroacetylacetone
CAS: 367-57-7; EC No: 206-698-5; MDL No: MFCD00000427
UN No: UN1224; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CF3COCH2COCH3; Molecular Formula: C5H5F3O2; MW: 154.09
Boiling Point: 105-107°; Flash point: 26°C (79°F)
Density (g/mL): 1.270; Refractive Index: 1.388
- Refractive index: 1.388
- Liquid
Formula:
C₅H₅F₃O₂ MW: 154.09 g/mol Boiling Pt: 105…107 °C Density: 1.270 g/mL Flash Pt: 26 °C (79 °F) |
MDL Number:
MFCD00000427 CAS Number: 367-57-7 EINECS: 206-698-5 UN: 1224 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...