(R)-(-)-2-Amino-1-propanol ≥98%
Supplier: BeanTown Chemical
Synonyms:
D-Alaninol, D(-)-2-Amino-1-propanol, D(-)-Alaninol, (R)-2-Aminopropan-1-ol
225840-1G
225840-25G
225840-5G
BT225840-1GEA
40
USD
BT225840-1G
BT225840-25G
BT225840-5G
(R)-(-)-2-Amino-1-propanol ≥98%
(R)-(-)-2-Amino-1-propanol
CAS: 35320-23-1; MDL No: MFCD00064413
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: CH3CH(NH2)CH2OH; Molecular Formula: C3H9NO; MW: 75.11
Boiling Point: 166-168°; Flash point: 83°C (181°F)
Density (g/mL): 0.963; Refractive Index: 1.449; Optical Rotation: [α]19/D −18°, neat
Air Sensitive , Hygroscopic
- Optical rotation: [α]19/D −18°, neat
- Refractive index: 1.449
- Air Sensitive, Hygroscopic
- Liquid
Formula:
C₃H₉NO Boiling Pt: 166…168 °C Density: 0.963 g/mL Flash Pt: 83 °C (181 °F) |
MDL Number:
MFCD00064413 CAS Number: 35320-23-1 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...