Methyltrioxorhenium(VII) ≥98%
Supplier: BeanTown Chemical
225825-100MG
225825-500MG
BT225825-100MGEA
41
USD
BT225825-100MG
BT225825-500MG
Methyltrioxorhenium(VII) ≥98%
Methyltrioxorhenium(VII)
CAS: 70197-13-6; MDL No: MFCD00192332
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: CH3ReO3; MW: 249.24
Melting Point: 110-115°
Light Sensitive, Moisture Sensitive
- Light Sensitive, Moisture Sensitive
- Crystalline
Formula:
CH₃O₃Re Melting Pt: 110…115 °C |
MDL Number:
MFCD00192332 CAS Number: 70197-13-6 ADR: 5.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...