Methacrylic anhydride ≥94% stabilized
Supplier: BeanTown Chemical
|
|
Synonyms:
Methacrylic Anhydride [stabilized with 1,1,3-Tris(3-tert-butyl-4-hydroxy-6-methylphenyl)butane]
225790-1KG
225790-250G
225790-50G
BT225790-1KGEA
365
USD
BT225790-1KG
BT225790-250G
BT225790-50G
Methacrylic anhydride ≥94% stabilized
Methacrylic anhydride
CAS: 760-93-0; EC No: 212-084-8; MDL No: MFCD00008586; RTECS: OZ5700000
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: [H2C=C(CH3)CO]2O; Molecular Formula: C8H10O3; MW: 154.17
Melting Point: -109°; Boiling Point: 197°; Flash point: 84°C (183°F)
Density (g/mL): 1.035; Refractive Index: 1.453
Light Sensitive, Moisture Sensitive
Formula:
C₈H₁₀O₃ MW: 154.165 g/mol Boiling Pt: 197 °C Melting Pt: –109 °C Density: 1.035 g/cm³ (25 °C) Flash Pt: 84 °C |
MDL Number:
MFCD00008586 CAS Number: 760-93-0 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 94% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...