Boc-ON ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-(tert-BOC-Oxyimino)-2-phenylacetonitrile, N-((tert-Butoxycarbonyl)oxy)benzimidoyl cyanide, 2-(Boc-oxyimino)-2-phenylacetonitrile, 2-(tert-Butoxycarbonyloxyimino)-2-phenylacetonitrile, [(cyano-phenyl-methylidene)amino] tert-butyl carbonate
225775-100G
225775-25G
225775-5G
BT225775-100GEA
506
USD
BT225775-100G
BT225775-25G
BT225775-5G
Boc-ON ≥98%
Boc-ON
CAS: 58632-95-4; EC No: 261-370-9; MDL No: MFCD00001863; RTECS: DA0513600
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C13H14N2O3; MW: 246.26
Melting Point: 85-88°
Moisture Sensitive
- RTECS: DA0513600
- Moisture Sensitive
- Crystalline
Formula:
C₁₃H₁₄N₂O₃ MW: 246.27 g/mol Melting Pt: 85…88 °C |
MDL Number:
MFCD00001863 CAS Number: 58632-95-4 EINECS: 261-370-9 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...