4-Methyl-1-pentyne ≥97%

Supplier: BeanTown Chemical

225765-1G 225765-5G
BT225765-1GEA 57 USD
BT225765-1G BT225765-5G
4-Methyl-1-pentyne ≥97%
4-Methyl-1-pentyne

CAS: 7154-75-8; MDL No: MFCD00039857 UN No: UN3295; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)2CHCH2C≡CH; Molecular Formula: C6H10; MW: 82.14 Boiling Point: 61-62°; Flash point: -4°C (25°F) Density (g/mL): 0.698; Refractive Index: 1.393


  • Refractive index: 1.393
  • Liquid

Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 61…62 °C
Density: 0.698 g/mL
Flash Pt: –4 °C (25 °F)
MDL Number: MFCD00039857
CAS Number: 7154-75-8
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR