2,2,4-Trimethylpentane ≥99.7% for HPLC

Supplier: BeanTown Chemical

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

225760-1L 225760-4L 225760-4X1L 225760-4X4L
BT225760-1LEA 80.85 USD
BT225760-1L BT225760-4L BT225760-4X1L BT225760-4X4L
2,2,4-Trimethylpentane ≥99.7% for HPLC
2,2,4-Trimethylpentane

CAS: 540-84-1; EC No: 208-759-1; MDL No: MFCD00008943; RTECS: SA3320000 UN No: UN1262; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)2CHCH2C(CH3)3; Molecular Formula: C8H18; MW: 114.23 Melting Point: -107°; Boiling Point: 98-99°; Flash point: -12°C (10°F) Density (g/mL): 0.692; Refractive Index: 1.391


  • RTECS: SA3320000
  • Refractive index: 1.391
  • Liquid

Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 98…99 °C
Melting Pt: –107 °C
Density: 0.692 g/mL
Flash Pt: –12 °C (10 °F)
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.7% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR