N-Chlorosuccinimide ≥98%
Supplier: BeanTown Chemical
Synonyms:
1-Chloropyrrolidine-2,5-dione
225750-1KG
225750-250G
225750-50G
BT225750-1KGEA
157.56
USD
BT225750-1KG
BT225750-250G
BT225750-50G
N-Chlorosuccinimide ≥98%
N-Chlorosuccinimide
CAS: 128-09-6; EC No: 204-878-8; MDL No: MFCD00005511; RTECS: UY1013500
UN No: UN3261; Haz Class: 8; Packing Group: II
Solid; Molecular Formula: C4H4ClNO2; MW: 133.53
Melting Point: 148-150°
Density (g/mL): 1.65
Light Sensitive, Moisture Sensitive
- RTECS: UY1013500
- Light Sensitive, Moisture Sensitive
- Solid
Formula:
C₄H₄ClNO₂ MW: 133.53 g/mol Melting Pt: 148…150 °C Density: 1.65 g/mL |
MDL Number:
MFCD00005511 CAS Number: 128-09-6 EINECS: 204-878-8 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...