2,3-Dichloro-5,6-dicyano-p-benzoquinone ≥98%
Supplier: BeanTown Chemical
Synonyms:
DDQ, 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 2,3-Dichloro-5,6-dicyanoquinone, 4,5-Dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
225720-10G
225720-250G
225720-50G
BT225720-10GEA
49.22
USD
BT225720-10G
BT225720-250G
BT225720-50G
2,3-Dichloro-5,6-dicyano-p-benzoquinone ≥98%
2,3-Dichloro-5,6-dicyano-p-benzoquinone
CAS: 84-58-2; EC No: 201-542-2; MDL No: MFCD00001593; RTECS: GU4825000
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C8Cl2N2O2; MW: 227.00
Melting Point: 210-215° (decomposes)
Moisture Sensitive
- RTECS: GU4825000
- Moisture Sensitive
- Crystalline
Formula:
C₈Cl₂N₂O₂ MW: 227.01 g/mol Melting Pt: 210…215 °C (dec.) |
MDL Number:
MFCD00001593 CAS Number: 84-58-2 EINECS: 201-542-2 UN: 3439 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...