Propionyl chloride ≥98%
Supplier: BeanTown Chemical
CAS: 79-03-8; EC No: 201-170-0; MDL No: MFCD00000745
UN No: UN1815; Haz Class: 3 (8); Packing Group: II
Liquid; Linear Formula: CH3CH2COCl; Molecular Formula: C3H5ClO; MW: 92.53
Melting Point: -94°; Boiling Point: 77-79°; Flash point: 6°C (43°F)
Density (g/mL): 1.059; Refractive Index: 1.404
Moisture Sensitive
- Refractive index: 1.404
- Moisture Sensitive
- Liquid
Formula:
C₃H₅ClO Boiling Pt: 77…79 °C Melting Pt: –94 °C Density: 1.059 g/mL Flash Pt: 6 °C (43 °F) |
MDL Number:
MFCD00000745 CAS Number: 79-03-8 EINECS: 201-170-0 UN: 1815 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...