Potassium hydrogen difluoride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Potassium hydrogen fluoride, Potassium bifluoride
225650-2KG
225650-500G
BT225650-2KGEA
210
USD
BT225650-2KG
BT225650-500G
Potassium hydrogen difluoride ≥99%
Potassium hydrogen difluoride
CAS: 7789-29-9; EC No: 232-156-2; MDL No: MFCD00011403; RTECS: TS6650000
UN No: UN1811; Haz Class: 8 (6.1); Packing Group: II
Crystalline; Linear Formula: KHF2; MW: 78.10
Melting Point: 239°
Density (g/mL): 2.37
Hygroscopic
- RTECS: TS6650000
- Hygroscopic
- Crystalline
Formula:
HF₂K MW: 78.1 g/mol Melting Pt: 239 Density: 2.37 g/mL |
MDL Number:
MFCD00011403 CAS Number: 7789-29-9 EINECS: 232-156-2 UN: 1811 ADR: 8 II Merck Index: 12,07771 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...